Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172.3 | NP_001163.1 | 3 | 440 | ||
GenBank mRNA | AK312484.1 | - | 3 | 356 | ||
AY074489.1 | - | 3 | 385 | |||
BC001350.1 | - | 3 | 341 | 2964483 | ||
BC008464.1 | - | 3 | 382 | 4274290 | ||
BC029050.1 | - | 3 | 440 | 5179867 | ||
CR536550.1 | - | 3 | 324 | |||
D86724.1 | - | 3 | 344 | |||
U75667.1 | - | 3 | 362 | |||
U82256.1 | - | 3 | 340 |