Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENTPD6 SIRNA Assays ›
Gene Symbol:
ENTPD6
Gene Name:
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases:
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
Chromosome Location:
Chr.20: 25195670 - 25228075 on Build GRCh38
UniGene ID:
Hs.500375
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119117
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ENTPD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114089.3
NP_001107561.1
14
2308
NM_001247.4
NP_001238.2
15
2377
NM_001317941.2
NP_001304870.1
15
2374
NM_001322378.1
NP_001309307.1
15
2521
NM_001322379.1
NP_001309308.1
12
1884
NM_001322380.1
NP_001309309.1
13
2029
NM_001322381.1
NP_001309310.1
12
1885
NM_001322382.1
NP_001309311.1
12
1842
NM_001322383.1
NP_001309312.1
16
2565
NM_001322384.1
NP_001309313.1
13
1986
NM_001322385.1
NP_001309314.1
13
2164
NM_001322386.1
NP_001309315.1
13
2206
NM_001322387.1
NP_001309316.1
14
2233
NM_001322388.1
NP_001309317.1
14
2318
NM_001322389.1
NP_001309318.1
13
2350
NM_001322390.1
NP_001309319.1
15
2397
NM_001322391.1
NP_001309320.1
14
2275
NM_001322392.1
NP_001309321.1
14
2348
NM_001322393.1
NP_001309322.1
14
2351
NM_001322394.1
NP_001309323.1
14
2305
NM_001322395.1
NP_001309324.1
14
2495
NM_001322396.1
NP_001309325.1
15
2417
NM_001322397.1
NP_001309326.1
15
2420
NM_001322398.1
NP_001309327.1
14
2452
XM_006723665.2
XP_006723728.1
16
2413
XM_011529396.1
XP_011527698.1
16
2417
XM_011529397.1
XP_011527699.1
16
2346
XM_017028129.1
XP_016883618.1
13
2127
XM_017028130.1
XP_016883619.1
13
2042
XM_017028132.1
XP_016883621.1
14
2209
XM_017028133.1
XP_016883622.1
13
2097
XM_017028134.1
XP_016883623.1
12
1894
GenBank mRNA
AF039916.1
-
15
2310
AK074691.1
-
14
2157
AK094030.1
-
4
2165
AK095708.1
-
2
1495
AK124625.1
-
4
3981
AK128412.1
-
9
2742
AL834158.1
-
14
2223
AY327581.1
-
15
2274
BC025980.2
-
15
2280
3636163
CR936750.1
-
13
1930
CR936765.1
-
15
2334
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
nucleotide phosphatase
phosphatase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
response to magnesium ion
response to calcium ion
guanosine-5'-triphosphate,3'-diphosphate diphosphatase activity
nucleoside-triphosphatase activity
uridine-diphosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline