Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001249.3 | NP_001240.1 | 4 | 397 | ||
NM_001321984.1 | NP_001308913.1 | 5 | 524 | |||
NM_001321985.1 | NP_001308914.1 | 5 | 514 | |||
NM_001321986.1 | NP_001308915.1 | 5 | 494 | |||
NM_001321987.1 | NP_001308916.1 | 6 | 641 | |||
NM_001321988.1 | NP_001308917.1 | 4 | 1173 | |||
XM_005268224.3 | XP_005268281.1 | 4 | 359 | |||
XM_006720325.3 | XP_006720388.1 | 4 | 400 | |||
XM_006720326.2 | XP_006720389.1 | 4 | 400 | |||
XM_017021813.1 | XP_016877302.1 | 5 | 519 | |||
XM_017021814.1 | XP_016877303.1 | 2 | 274 | |||
XM_017021815.1 | XP_016877304.1 | 5 | 519 | |||
XM_017021816.1 | XP_016877305.1 | 2 | 274 | |||
XM_017021817.1 | XP_016877306.1 | 4 | 400 | |||
GenBank mRNA | AF039918.1 | - | 3 | 332 | ||
AF136572.1 | - | 3 | 368 | |||
AY430094.1 | - | 3 | 338 | |||
BC130485.1 | - | 1 | 114 | 40146516 | ||
BC130487.1 | - | 1 | 114 | 40146518 |