Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197293.2 | NP_001184222.1 | 2 | 540 | ||
NM_001244604.1 | NP_001231533.1 | 2 | 182 | |||
NM_001386.5 | NP_001377.1 | 2 | 523 | |||
GenBank mRNA | AB209195.1 | - | 2 | 261 | ||
AB370194.1 | - | 2 | 100 | |||
AB370195.1 | - | 2 | 458 | |||
AK223557.1 | - | 2 | 386 | |||
AK291287.1 | - | 2 | 304 | |||
AK299077.1 | - | 2 | 175 | |||
BC056408.1 | - | 2 | 377 | 6044141 | ||
BC067109.1 | - | 2 | 407 | 3870039 | ||
BP222303.1 | - | 2 | 182 | |||
BX461557.2 | - | 2 | 370 | |||
BX463316.2 | - | 2 | 365 | |||
BX647115.1 | - | 2 | 451 | |||
D78013.1 | - | 2 | 366 | |||
DC333865.1 | - | 2 | 504 | |||
DC344356.1 | - | 2 | 521 | |||
U17279.1 | - | 2 | 163 |