Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197293.2 | NP_001184222.1 | 5, 6 | 981 | ||
NM_001244604.1 | NP_001231533.1 | 5, 6 | 623 | |||
NM_001386.5 | NP_001377.1 | 5, 6 | 964 | |||
GenBank mRNA | AB209195.1 | - | 5, 6 | 702 | ||
AK223557.1 | - | 5, 6 | 827 | |||
AK291287.1 | - | 5, 6 | 745 | |||
AK299077.1 | - | 5, 6 | 616 | |||
BC056408.1 | - | 5, 6 | 818 | 6044141 | ||
BC067109.1 | - | 5, 6 | 848 | 3870039 | ||
BX463316.2 | - | 5, 6 | 806 | |||
BX647115.1 | - | 5, 6 | 892 | |||
D78013.1 | - | 5, 6 | 807 | |||
U17279.1 | - | 5, 6 | 604 |