Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197294.1 | NP_001184223.1 | 3 | 791 | ||
NM_001387.2 | NP_001378.1 | 3 | 361 | |||
XM_011537574.1 | XP_011535876.1 | 4 | 745 | |||
GenBank mRNA | BC039006.1 | - | 3 | 327 | 5725550 | |
BC077077.1 | - | 3 | 791 | 6177053 | ||
D78014.1 | - | 3 | 361 | |||
EU007906.1 | - | 3 | 593 | |||
Y07818.1 | - | 3 | 382 |