Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197294.1 | NP_001184223.1 | 14 | 2326 | ||
NM_001387.2 | NP_001378.1 | 14 | 1896 | |||
XM_011537574.1 | XP_011535876.1 | 15 | 2280 | |||
GenBank mRNA | AK297223.1 | - | 5 | 771 | ||
BC039006.1 | - | 14 | 1862 | 5725550 | ||
BC077077.1 | - | 14 | 2326 | 6177053 | ||
D78014.1 | - | 14 | 1896 |