Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002641.3 | NP_002632.1 | 6 | 2317 | ||
NM_020473.3 | NP_065206.3 | 5 | 1540 | |||
NR_033835.1 | 6 | 1943 | ||||
NR_033836.1 | 6 | 1659 | ||||
XM_011545539.2 | XP_011543841.1 | 5 | 3141 | |||
XM_017029581.1 | XP_016885070.1 | 5 | 2264 | |||
GenBank mRNA | AK098047.1 | - | 5 | 1753 | ||
BC038236.1 | - | 5 | 2274 | 5733899 | ||
D11466.1 | - | 6 | 2286 |