Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001329.2 | NP_001001329.1 | 3 | 685 | ||
NM_001289102.1 | NP_001276031.1 | 2 | 536 | |||
NM_001289103.1 | NP_001276032.1 | 3 | 689 | |||
NM_001289104.1 | NP_001276033.1 | 3 | 685 | |||
NM_002743.3 | NP_002734.2 | 3 | 297 | |||
XM_011528130.1 | XP_011526432.1 | 4 | 451 | |||
XM_011528131.1 | XP_011526433.1 | 4 | 451 | |||
XM_011528132.1 | XP_011526434.1 | 4 | 451 | |||
XM_017026977.1 | XP_016882466.1 | 3 | 301 | |||
GenBank mRNA | AF144075.1 | - | 2 | 158 | ||
AK130663.1 | - | 3 | 313 | |||
AK225279.1 | - | 3 | 311 | |||
AK225390.1 | - | 3 | 293 | |||
AK290433.1 | - | 2 | 531 | |||
AK296187.1 | - | 3 | 312 | |||
BC013586.1 | - | 2 | 95 | |||
BG323695.1 | - | 508 | ||||
BI254784.1 | - | 3 | 493 | |||
BP242217.1 | - | 2 | 536 | |||
DB024838.1 | - | 3 | 391 | |||
J03075.1 | - | 3 | 294 | |||
Z36798.1 | - | 3 | 294 |