Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032940.2 | NP_116558.1 | 8, 9 | 763 | ||
GenBank mRNA | AF008443.1 | - | 8, 9 | 677 | ||
AJ224143.1 | - | 8, 9 | 716 | |||
AJ224144.1 | - | 8, 9 | 716 | |||
AK293112.1 | - | 8, 9 | 721 | |||
AK300124.1 | - | 734 | ||||
BC000409.2 | - | 8, 9 | 694 | 2960943 | ||
BC003159.1 | - | 8, 9 | 735 | 2957867 | ||
BC028157.1 | - | 8, 9 | 719 | 5240953 | ||
CR542041.1 | - | 8, 9 | 677 | |||
J05448.1 | - | 8, 9 | 718 |