Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127505.2 | NP_001120977.1 | 5 | 538 | ||
NM_004315.5 | NP_004306.3 | 5 | 604 | |||
NM_177924.4 | NP_808592.2 | 5 | 671 | |||
XM_005273504.3 | XP_005273561.1 | 5 | 432 | |||
GenBank mRNA | AK025732.1 | - | 6 | 574 | ||
AK098299.1 | - | 6 | 394 | |||
AK222780.1 | - | 6 | 407 | |||
AK289481.1 | - | 6 | 572 | |||
BC016481.1 | - | 6 | 394 | 3923451 | ||
BC016828.1 | - | 5 | 525 | 4109317 | ||
BG714023.1 | - | 5 | 418 | |||
U47674.1 | - | 5 | 375 | |||
U70063.1 | - | 6 | 376 |