Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079866.1 | NP_001073335.1 | 2 | 321 | ||
NM_001257342.1 | NP_001244271.1 | 3 | 451 | |||
NM_001257343.1 | NP_001244272.1 | 3 | 503 | |||
NM_001257344.1 | NP_001244273.1 | 2 | 378 | |||
NM_001320717.1 | NP_001307646.1 | 3 | 493 | |||
NM_004328.4 | NP_004319.1 | 3 | 530 | |||
XM_005246748.2 | XP_005246805.1 | 2 | 830 | |||
XM_006712678.1 | XP_006712741.1 | 2 | 936 | |||
XM_017004631.1 | XP_016860120.1 | 3 | 692 | |||
XM_017004632.1 | XP_016860121.1 | 3 | 417 | |||
XM_017004633.1 | XP_016860122.1 | 2 | 1220 | |||
XM_017004634.1 | XP_016860123.1 | 1 | 248 | |||
GenBank mRNA | AF026849.1 | - | 2 | 297 | ||
AF038195.1 | - | 2 | 259 | |||
AK096210.1 | - | 2 | 884 | |||
AK223518.1 | - | 2 | 407 | |||
AK289363.1 | - | 3 | 393 | |||
BC000416.2 | - | 2 | 451 | 2961496 | ||
BC007500.2 | - | 2 | 451 | 2961496 | ||
BX571752.1 | - | 3 | 450 | |||
CB159298.1 | - | 3 | 420 | |||
DA540458.1 | - | 2 | 321 | |||
DA613959.1 | - | 2 | 321 | |||
KP992846.1 | - | 3 | 450 | |||
KU177941.1 | - | 1 | 184 | |||
KU177942.1 | - | 1 | 184 |