Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302777.1 | NP_001289706.1 | 5, 6 | 1119 | ||
NM_004332.3 | NP_004323.2 | 4, 5 | 891 | |||
NR_026648.2 | 5, 6 | 1119 | ||||
NR_026649.2 | 4, 5 | 891 | ||||
GenBank mRNA | AJ617684.1 | - | 4, 5 | 539 | ||
AK297518.1 | - | 4, 5 | 575 | |||
AK314909.1 | - | 5, 6 | 773 | |||
AL832502.1 | - | 4, 5 | 542 | |||
BC037778.1 | - | 5, 6 | 1124 | 4799692 | ||
BC106901.1 | - | 4, 5 | 606 | 40031474 | ||
X81372.1 | - | 5, 6 | 687 |