Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004665.4 | NP_004656.2 | 5 | 1204 | ||
NM_078488.2 | NP_511043.1 | 6 | 1032 | |||
NR_034173.2 | 4 | 915 | ||||
NR_034174.2 | 4 | 675 | ||||
NR_110143.1 | 5 | 1200 | ||||
NR_110144.1 | 5 | 992 | ||||
NR_110145.1 | 5 | 989 | ||||
XM_006715593.3 | XP_006715656.1 | 7 | 1179 | |||
XM_011536231.1 | XP_011534533.1 | 5 | 1100 | |||
XM_017011407.1 | XP_016866896.1 | 6 | 1815 | |||
XM_017011408.1 | XP_016866897.1 | 7 | 1184 | |||
XM_017011409.1 | XP_016866898.1 | 7 | 1077 | |||
GenBank mRNA | AB026705.1 | - | 5 | 1086 | ||
AB435062.1 | - | 4 | 790 | |||
AB435063.1 | - | 4 | 550 | |||
AJ132100.1 | - | 5 | 1090 | |||
AK290908.1 | - | 5 | 1100 | |||
AK292125.1 | - | 6 | 1020 | |||
AK298719.1 | - | 5 | 885 | |||
BC126145.1 | - | 5 | 1166 | 8991861 | ||
BC126147.1 | - | 5 | 1166 | 8991863 | ||
D89974.1 | - | 5 | 1090 |