Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005908.3 | NP_005899.3 | 5 | 733 | ||
XM_017008203.1 | XP_016863692.1 | 6 | 719 | |||
GenBank mRNA | AB209846.1 | - | 5 | 680 | ||
AK291598.1 | - | 5 | 721 | |||
AK300015.1 | - | 4 | 503 | |||
AK312827.1 | - | 5 | 688 | |||
BC015743.1 | - | 5 | 723 | 4905535 | ||
EU009130.1 | - | 5 | 633 | |||
U60337.1 | - | 5 | 720 |