Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098531.2 | NP_001092001.1 | 11, 12 | 1706 | ||
NM_001098532.2 | NP_001092002.1 | 10, 11 | 1470 | |||
NM_006105.5 | NP_006096.2 | 11, 12 | 1240 | |||
XM_005268571.3 | XP_005268628.1 | 10, 11 | 1250 | |||
XM_011537752.1 | XP_011536054.1 | 11, 12 | 1723 | |||
XM_011537755.1 | XP_011536057.1 | 11, 12 | 1285 | |||
XM_011537758.2 | XP_011536060.1 | 11, 12 | 1335 | |||
XM_017018688.1 | XP_016874177.1 | 11, 12 | 1723 | |||
GenBank mRNA | AF103905.1 | - | 10, 11 | 1030 | ||
AK092448.1 | - | 11, 12 | 1678 | |||
AK125545.1 | - | 7, 8 | 723 | |||
AK290230.1 | - | 12, 13 | 1371 | |||
AK299029.1 | - | 11, 12 | 1334 | |||
AK308079.1 | - | 11, 12 | 1308 | |||
BC017728.1 | - | 11, 12 | 1320 | 4414360 | ||
BC092404.1 | - | 11, 12 | 1309 | 6181000 | ||
CN294139.1 | - | 6, 7 | 602 | |||
U78168.1 | - | 11, 12 | 1245 | |||
U78169.1 | - | 11, 12 | 1245 |