Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006426.2 | NP_006417.2 | 14 | 2491 | ||
XM_005252658.3 | XP_005252715.1 | 14 | 2480 | |||
XM_011539125.2 | XP_011537427.1 | 15 | 2494 | |||
XM_017015482.1 | XP_016870971.1 | 14 | 3755 | |||
XM_017015483.1 | XP_016870972.1 | 15 | 3908 | |||
GenBank mRNA | AB006713.1 | - | 14 | 2485 |