Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134445.1 | NP_001127917.1 | 7 | 1834 | ||
NM_012137.3 | NP_036269.1 | 6 | 1802 | |||
XM_005270707.2 | XP_005270764.1 | 6 | 1511 | |||
XM_005270710.2 | XP_005270767.1 | 6 | 1730 | |||
XM_011541158.1 | XP_011539460.1 | 8 | 1834 | |||
XM_017000889.1 | XP_016856378.1 | 6 | 1488 | |||
XM_017000890.1 | XP_016856379.1 | 6 | 3240 | |||
GenBank mRNA | AK024917.1 | - | 1 | 249 | ||
AK098405.1 | - | 7 | 710 | |||
AW051386.1 | - | 1 | 402 | |||
BC036432.2 | - | 1 | 59 | |||
BC043235.2 | - | 6 | 1525 | |||
BX648145.1 | - | 7 | 1718 |