Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002775.4 | NP_002766.1 | 1, 2 | 608 | ||
GenBank mRNA | AF097709.1 | - | 1, 2 | 51 | ||
AK091944.1 | - | 3, 4 | 383 | |||
AK092476.1 | - | 2, 3 | 389 | |||
BC011352.1 | - | 2, 3 | 375 | |||
BC031082.1 | - | 1, 2 | 516 | |||
D87258.1 | - | 1, 2 | 528 | |||
DA633502.1 | - | 3, 4 | 383 | |||
Y07921.1 | - | 1, 2 | 518 |