Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013358.2 | NP_037490.2 | 16 | 2450 | ||
XM_017001101.1 | XP_016856590.1 | 16 | 2353 | |||
XM_017001102.1 | XP_016856591.1 | 16 | 2353 | |||
GenBank mRNA | AB033768.2 | - | 16 | 2441 | ||
AK026652.1 | - | 16 | 2452 | |||
AK127515.1 | - | 2 | 1729 | |||
AK298538.1 | - | 5 | 1075 | |||
AK298892.1 | - | 5 | 984 | |||
AK309106.1 | - | 2 | 689 |