Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042402.1 | NP_001035861.1 | 2 | 303 | ||
NM_014435.3 | NP_055250.2 | 2 | 303 | |||
XM_005262920.3 | XP_005262977.1 | 2 | 301 | |||
XM_005262921.3 | XP_005262978.1 | 2 | 301 | |||
XM_005262923.2 | XP_005262980.1 | 2 | 301 | |||
XM_005262924.2 | XP_005262981.1 | 2 | 301 | |||
XM_005262925.3 | XP_005262982.1 | 2 | 301 | |||
XM_006714180.2 | XP_006714243.1 | 2 | 301 | |||
XM_011531852.2 | XP_011530154.1 | 2 | 301 | |||
XM_011531854.2 | XP_011530156.1 | 2 | 301 | |||
XM_017008027.1 | XP_016863516.1 | 2 | 301 | |||
XM_017008028.1 | XP_016863517.1 | 2 | 301 | |||
XM_017008029.1 | XP_016863518.1 | 2 | 301 | |||
XM_017008030.1 | XP_016863519.1 | 2 | 301 | |||
GenBank mRNA | AB161353.1 | - | 2 | 239 | ||
AK024677.1 | - | 803 | ||||
AK301132.1 | - | 1 | 128 | |||
AK309271.1 | - | 2 | 275 | |||
BC006388.2 | - | 2 | 283 | 4122755 | ||
BM763257.1 | - | 2 | 303 |