Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198665.1 | NP_001185594.1 | 3 | 454 | ||
NM_001301084.1 | NP_001288013.1 | 3 | 280 | |||
NM_015313.2 | NP_056128.1 | 3 | 454 | |||
XM_006718805.3 | XP_006718868.1 | 3 | 813 | |||
XM_011542720.2 | XP_011541022.1 | 3 | 395 | |||
XM_017017420.1 | XP_016872909.1 | 3 | 1087 | |||
XM_017017421.1 | XP_016872910.1 | 3 | 670 | |||
XM_017017422.1 | XP_016872911.1 | 3 | 282 | |||
GenBank mRNA | AF180681.1 | - | 3 | 126 | ||
AK294803.1 | - | 3 | 255 | |||
AK304316.1 | - | 4 | 735 | |||
AL137456.1 | - | 2 | 273 | |||
BC063117.1 | - | 3 | 457 | |||
BX647785.1 | - | 3 | 256 | |||
DC350360.1 | - | 3 | 280 |