Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198665.1 | NP_001185594.1 | 25 | 2767 | ||
NM_001301084.1 | NP_001288013.1 | 26 | 2650 | |||
NM_015313.2 | NP_056128.1 | 26 | 2824 | |||
XM_006718805.3 | XP_006718868.1 | 25 | 3108 | |||
XM_011542720.2 | XP_011541022.1 | 26 | 2765 | |||
XM_017017420.1 | XP_016872909.1 | 24 | 3280 | |||
XM_017017421.1 | XP_016872910.1 | 26 | 3040 | |||
XM_017017422.1 | XP_016872911.1 | 25 | 2532 | |||
GenBank mRNA | AB002380.2 | - | 21 | 2186 | ||
AF180681.1 | - | 26 | 2496 | |||
AK294803.1 | - | 26 | 2625 | |||
AK304316.1 | - | 26 | 3048 | |||
BX647785.1 | - | 26 | 2626 |