Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145815.1 | NP_001139287.1 | 3 | 448 | ||
NM_015944.3 | NP_057028.2 | 3 | 448 | |||
XM_017023262.1 | XP_016878751.1 | 3 | 459 | |||
XM_017023263.1 | XP_016878752.1 | 3 | 459 | |||
XM_017023264.1 | XP_016878753.1 | 3 | 459 | |||
XM_017023265.1 | XP_016878754.1 | 3 | 459 | |||
XM_017023266.1 | XP_016878755.1 | 3 | 407 | |||
XM_017023267.1 | XP_016878756.1 | 2 | 311 | |||
GenBank mRNA | AK296877.1 | - | 3 | 420 | ||
AK300747.1 | - | 2 | 264 | |||
BC018734.1 | - | 3 | 384 | 4869721 | ||
CN362870.1 | - | 2 | 318 |