Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270965.1 | NP_001257894.1 | 5 | 1309 | ||
NM_001270966.1 | NP_001257895.1 | 5 | 830 | |||
NM_002779.4 | NP_002770.3 | 4 | 1633 | |||
XM_011539968.1 | XP_011538270.1 | 5 | 1315 | |||
XM_011539969.2 | XP_011538271.1 | 5 | 1332 | |||
XM_017016433.1 | XP_016871922.1 | 5 | 1326 | |||
XM_017016434.1 | XP_016871923.1 | 4 | 1253 | |||
GenBank mRNA | AB095931.1 | - | 4 | 1633 | ||
BC039259.1 | - | 5 | 817 | |||
BC142643.1 | - | 5 | 1309 | 40147927 | ||
BC142689.1 | - | 5 | 1309 | 40148656 |