Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303533.1 | NP_001290462.1 | 7 | 741 | ||
NM_001303534.1 | NP_001290463.1 | 7 | 836 | |||
NM_016339.4 | NP_057423.1 | 7 | 1021 | |||
XM_006721932.3 | XP_006721995.1 | 7 | 1515 | |||
XM_006721935.3 | XP_006721998.1 | 8 | 792 | |||
XM_006721937.3 | XP_006722000.1 | 7 | 2617 | |||
XM_006721938.3 | XP_006722001.1 | 4 | 408 | |||
GenBank mRNA | AF117946.1 | - | 7 | 656 | ||
AK294642.1 | - | 3 | 643 | |||
AK304160.1 | - | 7 | 836 | |||
AK313492.1 | - | 7 | 696 | |||
BC029956.1 | - | 7 | 1515 | |||
BC115374.1 | - | 5 | 550 | 40028278 | ||
BC115375.1 | - | 5 | 550 | 40028280 |