Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017582.6 | NP_060052.3 | 8 | 1030 | ||
GenBank mRNA | AJ243666.1 | - | 2 | 97 | ||
AK056388.1 | - | 4 | 330 | |||
AK293986.1 | - | 8 | 653 | |||
AY112698.1 | - | 8 | 1005 | |||
AY948200.1 | - | 8 | 1013 | |||
BC000848.1 | - | 4 | 364 | |||
BC009286.2 | - | 3 | 168 | |||
BC061583.1 | - | 4 | 342 | |||
BC070158.1 | - | 7 | 555 | |||
BC087836.1 | - | 6 | 441 | |||
BC107057.2 | - | 9 | 698 | |||
BC148185.2 | - | 8 | 707 |