Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032020.4 | NP_114409.2 | 3 | 893 | ||
GenBank mRNA | AK075458.1 | - | 3 | 699 | ||
AK315475.1 | - | 3 | 745 | |||
AY358551.1 | - | 3 | 710 | |||
BC003060.2 | - | 3 | 674 | 3505533 | ||
BC051268.1 | - | 3 | 728 | 4855296 |