Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321967.1 | NP_001308896.1 | 5 | 537 | ||
NM_001321968.1 | NP_001308897.1 | 5 | 537 | |||
NM_001321969.1 | NP_001308898.1 | 6 | 627 | |||
NM_032810.3 | NP_116199.2 | 5 | 909 | |||
NR_135914.1 | 5 | 537 | ||||
XM_005270252.4 | XP_005270309.1 | 5 | 631 | |||
XM_011540302.1 | XP_011538604.1 | 3 | 302 | |||
XM_011540303.2 | XP_011538605.1 | 5 | 584 | |||
XM_017016847.1 | XP_016872336.1 | 5 | 778 | |||
XM_017016848.1 | XP_016872337.1 | 5 | 871 | |||
XM_017016849.1 | XP_016872338.1 | 6 | 750 | |||
XM_017016850.1 | XP_016872339.1 | 6 | 801 | |||
XM_017016851.1 | XP_016872340.1 | 5 | 584 | |||
GenBank mRNA | AF361493.1 | - | 4 | 743 | ||
AK027506.1 | - | 2 | 137 | |||
AK075223.1 | - | 5 | 838 | |||
AK304297.1 | - | 3 | 300 | |||
AL834370.1 | - | 4 | 322 | |||
BC063530.1 | - | 4 | 368 | |||
BC073998.1 | - | 4 | 480 | 6199813 |