Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321967.1 | NP_001308896.1 | 8 | 885 | ||
NM_001321968.1 | NP_001308897.1 | 7 | 795 | |||
NM_001321969.1 | NP_001308898.1 | 9 | 975 | |||
NM_032810.3 | NP_116199.2 | 8 | 1257 | |||
NR_135914.1 | 8 | 885 | ||||
XM_005270252.4 | XP_005270309.1 | 8 | 979 | |||
XM_011540302.1 | XP_011538604.1 | 6 | 650 | |||
XM_011540303.2 | XP_011538605.1 | 8 | 932 | |||
XM_017016847.1 | XP_016872336.1 | 8 | 1126 | |||
XM_017016848.1 | XP_016872337.1 | 7 | 1129 | |||
XM_017016849.1 | XP_016872338.1 | 9 | 1098 | |||
XM_017016850.1 | XP_016872339.1 | 9 | 1149 | |||
GenBank mRNA | AF361493.1 | - | 7 | 1091 | ||
AK027506.1 | - | 5 | 485 | |||
AK075223.1 | - | 8 | 1186 | |||
AK304297.1 | - | 5 | 558 | |||
AL834370.1 | - | 7 | 670 | |||
BC010868.1 | - | 4 | 320 | |||
BC063530.1 | - | 7 | 716 | |||
BC073998.1 | - | 7 | 828 | 6199813 |