Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138340.4 | NP_612213.2 | 5, 6 | 815 | ||
XM_011525847.2 | XP_011524149.1 | 5, 6 | 788 | |||
GenBank mRNA | AF007152.1 | - | 5, 6 | 774 | ||
AK024880.1 | - | 5, 6 | 689 | |||
BC021196.2 | - | 5, 6 | 775 | 3942164 |