Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004706.3 | NP_004697.2 | 23, 24 | 2358 | ||
NM_198977.1 | NP_945328.1 | 22, 23 | 2259 | |||
NM_199002.1 | NP_945353.1 | 23, 24 | 2383 | |||
XM_005259386.4 | XP_005259443.1 | 25, 26 | 2890 | |||
XM_005259390.4 | XP_005259447.1 | 23, 24 | 2706 | |||
XM_011527468.2 | XP_011525770.1 | 24, 25 | 2718 | |||
GenBank mRNA | AL832294.1 | - | 14, 15 | 1522 | ||
BC005155.2 | - | 20, 21 | 2096 | |||
BC011726.2 | - | 22, 23 | 2234 | 4138058 | ||
BC034013.1 | - | 23, 24 | 2314 | 3451036 | ||
BC035127.1 | - | 10, 11 | 1947 | |||
BC067262.1 | - | 22, 23 | 2202 | 6502165 | ||
BT007421.1 | - | 20, 21 | 2072 | |||
U64105.1 | - | 23, 24 | 2303 |