Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000263.3 | NP_000254.2 | 5 | 1242 | ||
XM_006721920.2 | XP_006721983.1 | 5 | 546 | |||
XM_017024686.1 | XP_016880175.1 | 4 | 775 | |||
XM_017024687.1 | XP_016880176.1 | 4 | 890 | |||
GenBank mRNA | BC053991.1 | - | 5 | 925 | 6213852 | |
L78464.1 | - | 5 | 908 | |||
U40846.1 | - | 5 | 1003 | |||
U43573.1 | - | 5 | 902 |