Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001064.3 | NP_001055.1 | 5 | 692 | ||
NM_001135055.2 | NP_001128527.1 | 5 | 692 | |||
NM_001258028.1 | NP_001244957.1 | 6 | 716 | |||
XM_011534054.1 | XP_011532356.1 | 6 | 654 | |||
XM_011534055.1 | XP_011532357.1 | 6 | 741 | |||
GenBank mRNA | AK093667.1 | - | 4 | 351 | ||
AK223614.1 | - | 5 | 600 | |||
AK289454.1 | - | 5 | 568 | |||
AK293438.1 | - | 6 | 624 | |||
AK303191.1 | - | 6 | 559 | |||
AK309482.1 | - | 7 | 730 | |||
BC002433.2 | - | 6 | 481 | 3346980 | ||
BC009970.2 | - | 5 | 590 | 4310396 | ||
BC024026.2 | - | 3 | 261 | |||
BX647248.1 | - | 5 | 578 | |||
BX649193.1 | - | 1 | 516 | |||
EU794667.1 | - | 5 | 590 | |||
KU178361.1 | - | 5 | 520 | |||
KU178362.1 | - | 5 | 520 | |||
L12711.1 | - | 5 | 618 | |||
U55017.1 | - | 5 | 520 | |||
X67688.1 | - | 6 | 590 |