Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320918.1 | NP_001307847.1 | 6 | 697 | ||
NM_001786.4 | NP_001777.1 | 6 | 685 | |||
NM_033379.4 | NP_203698.1 | 5 | 514 | |||
XM_005270303.3 | XP_005270360.1 | 6 | 767 | |||
GenBank mRNA | AK291939.1 | - | 6 | 622 | ||
AW976143.1 | - | 5 | 429 | |||
BC014563.1 | - | 5 | 556 | 2967728 | ||
BC107750.1 | - | 6 | 646 | 6710672 | ||
BT007004.1 | - | 5 | 543 | |||
CR933728.1 | - | 5 | 2912 | |||
D88357.1 | - | 4 | 399 | |||
X05360.1 | - | 6 | 669 | |||
Y00272.1 | - | 6 | 669 |