Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NVL SIRNA Assays ›
Gene Symbol:
NVL
Gene Name:
nuclear VCP-like
Gene Aliases:
NVL2
Chromosome Location:
Chr.1: 224227334 - 224330201 on Build GRCh38
UniGene ID:
Hs.497867
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119450
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NVL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243146.1
NP_001230075.1
2, 3
170
NM_001243147.1
NP_001230076.1
3, 4
244
NM_002533.3
NP_002524.2
3, 4
244
NM_206840.2
NP_996671.1
2, 3
170
XM_011544196.1
XP_011542498.1
3, 4
227
XM_011544198.1
XP_011542500.1
2, 3
182
XM_011544199.1
XP_011542501.1
3, 4
194
XM_011544200.1
XP_011542502.1
2, 3
168
XM_017001378.1
XP_016856867.1
3, 4
234
XM_017001379.1
XP_016856868.1
3, 4
234
XM_017001380.1
XP_016856869.1
3, 4
234
XM_017001381.1
XP_016856870.1
3, 4
227
XM_017001382.1
XP_016856871.1
2, 3
160
XM_017001383.1
XP_016856872.1
2, 3
160
XM_017001384.1
XP_016856873.1
3, 4
234
XM_017001385.1
XP_016856874.1
3, 4
234
XM_017001386.1
XP_016856875.1
2, 3
168
GenBank mRNA
AF401208.1
-
1, 2
51
AK293923.1
-
3, 4
203
AK297070.1
-
3, 4
203
AK297396.1
-
2, 3
121
AK298244.1
-
3, 4
193
AK298826.1
-
2, 3
122
BC005897.1
-
3, 4
203
BC012105.1
-
2, 3
102
4636004
BP284587.1
-
3, 4
244
U68140.1
-
3, 4
238
U78772.1
-
2, 3
151
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026171, AK013982, AK018049, BC031847, AK004676, and others.
Gene Ontology Categories:
Function(s)
Process(es)
ribosome biogenesis
positive regulation of telomerase activity
protein binding
ATP binding
poly(A) RNA binding
preribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-zrdb5:80/100.66.79.80:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline