Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003132.2 | NP_003123.2 | 3, 4 | 477 | ||
XM_017002179.1 | XP_016857668.1 | 3, 4 | 496 | |||
GenBank mRNA | BC000309.2 | - | 3, 4 | 443 | 2820975 | |
BC033106.1 | - | 3, 4 | 409 | 5420683 | ||
M34338.1 | - | 3, 4 | 468 |