Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040427.1 | NP_001035517.1 | 8, 9 | 1258 | ||
NM_001286249.1 | NP_001273178.1 | 6, 7 | 1053 | |||
NM_005326.4 | NP_005317.2 | 7, 8 | 1162 | |||
XM_011522469.2 | XP_011520771.1 | 7, 8 | 802 | |||
XM_011522470.2 | XP_011520772.1 | 6, 7 | 691 | |||
XM_017023195.1 | XP_016878684.1 | 7, 8 | 727 | |||
GenBank mRNA | AK290155.1 | - | 7, 8 | 727 | ||
AK298174.1 | - | 6, 7 | 693 | |||
AK299173.1 | - | 7, 8 | 793 | |||
AK299993.1 | - | 7, 8 | 793 | |||
BC000840.1 | - | 7, 8 | 782 | 3457214 | ||
BC002627.2 | - | 7, 8 | 1111 | 3608098 | ||
X90999.1 | - | 6, 7 | 647 |