Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286796.1 | NP_001273725.1 | 2 | 473 | ||
NM_001286797.1 | NP_001273726.1 | 2 | 286 | |||
NM_001286798.1 | NP_001273727.1 | 2 | 347 | |||
NM_001286799.1 | NP_001273728.1 | 2 | 276 | |||
NM_001286800.1 | NP_001273729.1 | 2 | 321 | |||
NM_001286801.1 | NP_001273730.1 | 2 | 347 | |||
NM_014064.3 | NP_054783.2 | 2 | 321 | |||
GenBank mRNA | AF110776.1 | - | 2 | 264 | ||
AK290957.1 | - | 2 | 318 | |||
AK292332.1 | - | 2 | 323 | |||
AK298840.1 | - | 2 | 311 | |||
AK313829.1 | - | 2 | 311 | |||
BC001396.2 | - | 2 | 300 | 3050940 | ||
BC033234.1 | - | 2 | 275 | 5426447 | ||
BE392289.1 | - | 2 | 278 | |||
BF316784.1 | - | 317 | ||||
BG468453.1 | - | 2 | 483 | |||
BI083998.1 | - | 2 | 286 | |||
BM554138.1 | - | 2 | 276 | |||
BQ923165.1 | - | 2 | 300 | |||
CN406947.1 | - | 2 | 334 |