Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039702.2 | NP_001034791.1 | 4 | 470 | ||
NM_018324.2 | NP_060794.1 | 5 | 629 | |||
XM_017016376.1 | XP_016871865.1 | 5 | 593 | |||
GenBank mRNA | AK001844.1 | - | 4 | 420 | ||
AK001968.1 | - | 5 | 562 | |||
BC050372.1 | - | 4 | 437 | 5742377 | ||
BG722790.1 | - | 4 | 472 |