Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018060.2 | NP_001018070.1 | 3 | 448 | ||
NM_001146288.1 | NP_001139760.1 | 3 | 533 | |||
NM_144704.2 | NP_653305.1 | 3 | 448 | |||
NR_027464.1 | 3 | 515 | ||||
GenBank mRNA | AK055035.1 | - | 3 | 432 | ||
AK094844.1 | - | 3 | 437 | |||
AK295581.1 | - | 3 | 418 | |||
AK316042.1 | - | 3 | 411 | |||
BC032485.1 | - | 3 | 415 | 5221203 | ||
CR456342.1 | - | 2 | 267 | |||
DA396585.1 | - | 3 | 470 | |||
DB172173.1 | - | 3 | 515 |