Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278346.1 | NP_001265275.1 | 2 | 1218 | ||
NM_153373.3 | NP_699204.1 | 2 | 395 | |||
NR_103508.1 | 2 | 395 | ||||
NR_103510.1 | 3 | 593 | ||||
XM_006714925.2 | XP_006714988.1 | 2 | 357 | |||
XM_011534669.2 | XP_011532971.1 | 2 | 357 | |||
XM_011534671.2 | XP_011532973.1 | 2 | 357 | |||
XM_011534672.2 | XP_011532974.1 | 2 | 143 | |||
XM_011534674.2 | XP_011532976.1 | 2 | 357 | |||
XM_017009995.1 | XP_016865484.1 | 2 | 357 | |||
XM_017009996.1 | XP_016865485.1 | 2 | 357 | |||
XM_017009997.1 | XP_016865486.1 | 2 | 357 | |||
XM_017009998.1 | XP_016865487.1 | 2 | 815 | |||
XM_017009999.1 | XP_016865488.1 | 1 | 960 | |||
GenBank mRNA | AK024076.1 | - | 1 | 1156 | ||
AK090397.1 | - | 1 | 707 | |||
AK292061.1 | - | 2 | 357 | |||
BC008009.1 | - | 2 | 363 | |||
BC037567.1 | - | 2 | 357 | 5220434 | ||
BC110335.1 | - | 2 | 367 | 5753177 | ||
BX417194.2 | - | 2 | 447 | |||
DB022121.1 | - | 3 | 391 |