Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290137.1 | NP_001277066.1 | 7 | 765 | ||
NM_001290139.1 | NP_001277068.1 | 7 | 835 | |||
NM_001323635.1 | NP_001310564.1 | 7 | 870 | |||
NM_001323636.1 | NP_001310565.1 | 7 | 870 | |||
NM_032304.3 | NP_115680.1 | 6 | 796 | |||
XM_005255631.4 | XP_005255688.1 | 7 | 772 | |||
XM_005255632.1 | XP_005255689.1 | 6 | 1030 | |||
XM_011522711.1 | XP_011521013.1 | 7 | 700 | |||
XM_011522712.1 | XP_011521014.1 | 6 | 1027 | |||
XM_017023773.1 | XP_016879262.1 | 7 | 660 | |||
XM_017023774.1 | XP_016879263.1 | 7 | 660 | |||
GenBank mRNA | AB209331.1 | - | 6 | 911 | ||
AK054841.1 | - | 6 | 779 | |||
AK098340.1 | - | 2 | 1424 | |||
AK293575.1 | - | 6 | 796 | |||
BC004353.1 | - | 6 | 771 | 3351990 | ||
BC015008.1 | - | 6 | 724 | 4862387 | ||
BG821643.1 | - | 7 | 582 | |||
BI333301.1 | - | 6 | 632 | |||
BI834617.1 | - | 2 | 101 | |||
CD050886.1 | - | 7 | 689 |