Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002802.2 | NP_002793.2 | 4 | 302 | ||
XM_017021470.1 | XP_016876959.1 | 4 | 1821 | |||
XM_017021471.1 | XP_016876960.1 | 5 | 378 | |||
GenBank mRNA | AB451311.1 | - | 1 | 254 | ||
AB451441.1 | - | 1 | 254 | |||
AK222668.1 | - | 4 | 270 | |||
AK299121.1 | - | 3 | 178 | |||
AK313325.1 | - | 4 | 291 | |||
BC000512.2 | - | 1 | 288 | 2822718 | ||
BC016368.1 | - | 1 | 282 | 4133348 | ||
BC018859.2 | - | 1 | 255 | |||
BC067741.1 | - | 1 | 275 | 5264945 | ||
BC073818.1 | - | 1 | 260 | 5456334 | ||
BT009826.1 | - | 1 | 254 | |||
CR457044.1 | - | 1 | 254 | |||
L02426.1 | - | 4 | 314 |