Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000310.3 | NP_000301.1 | 9 | 2222 | ||
NM_001142604.1 | NP_001136076.1 | 6 | 1913 | |||
XM_005271008.1 | XP_005271065.1 | 8 | 1934 | |||
GenBank mRNA | AK075496.1 | - | 4 | 1236 | ||
BC008426.1 | - | 9 | 2002 | 4249991 | ||
U44772.1 | - | 9 | 2007 |