Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000855.2 | NP_000846.1 | 8 | 2400 | ||
NM_001256424.1 | NP_001243353.1 | 9 | 2493 | |||
GenBank mRNA | BC130484.1 | - | 8 | 2106 | 40146515 | |
BC130488.1 | - | 8 | 2106 | 40146519 | ||
BC144033.1 | - | 9 | 2169 | 9052553 | ||
X63282.1 | - | 8 | 2400 | |||
Z50053.1 | - | 9 | 2493 |