Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000856.5 | NP_000847.2 | 11 | 2894 | ||
NM_001130682.2 | NP_001124154.1 | 10 | 2800 | |||
NM_001130683.3 | NP_001124155.1 | 10 | 2768 | |||
NM_001130684.2 | NP_001124156.1 | 9 | 2740 | |||
NM_001130685.2 | NP_001124157.1 | 10 | 2589 | |||
NM_001256449.1 | NP_001243378.1 | 10 | 2731 | |||
XM_005262955.2 | XP_005263012.1 | 10 | 2762 | |||
XM_005262956.2 | XP_005263013.1 | 11 | 2577 | |||
XM_005262957.2 | XP_005263014.1 | 11 | 2682 | |||
XM_006714196.2 | XP_006714259.1 | 10 | 2749 | |||
XM_006714197.2 | XP_006714260.1 | 10 | 2524 | |||
XM_006714198.2 | XP_006714261.1 | 10 | 2409 | |||
XM_011531900.2 | XP_011530202.1 | 10 | 2488 | |||
GenBank mRNA | AK226125.1 | - | 10 | 2457 | ||
BX647200.1 | - | 10 | 2561 | |||
BX649180.1 | - | 10 | 2523 | |||
X66534.1 | - | 11 | 2864 | |||
Y15723.1 | - | 11 | 2876 |