Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000857.3 | NP_000848.1 | 5 | 557 | ||
NM_001291951.1 | NP_001278880.1 | 6 | 623 | |||
NM_001291952.1 | NP_001278881.1 | 6 | 709 | |||
NM_001291953.1 | NP_001278882.1 | 6 | 743 | |||
NM_001291954.1 | NP_001278883.1 | 5 | 557 | |||
NM_001291955.1 | NP_001278884.1 | 4 | 483 | |||
XM_011531901.1 | XP_011530203.1 | 5 | 471 | |||
XM_017008130.1 | XP_016863619.1 | 5 | 748 | |||
XM_017008131.1 | XP_016863620.1 | 4 | 459 | |||
XM_017008132.1 | XP_016863621.1 | 5 | 748 | |||
XM_017008133.1 | XP_016863622.1 | 4 | 535 | |||
GenBank mRNA | AF020340.1 | - | 5 | 440 | ||
AK296680.1 | - | 6 | 656 | |||
AK299916.1 | - | 5 | 469 | |||
AK300296.1 | - | 6 | 575 | |||
AK307838.1 | - | 6 | 685 | |||
AK315199.1 | - | 5 | 510 | |||
AK315944.1 | - | 4 | 464 | |||
BC047620.1 | - | 5 | 471 | 5264160 | ||
X66533.1 | - | 5 | 477 |