Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000857.3 | NP_000848.1 | 8 | 1122 | ||
NM_001291951.1 | NP_001278880.1 | 9 | 1188 | |||
NM_001291952.1 | NP_001278881.1 | 9 | 1274 | |||
NM_001291953.1 | NP_001278882.1 | 9 | 1308 | |||
NM_001291954.1 | NP_001278883.1 | 8 | 1122 | |||
NM_001291955.1 | NP_001278884.1 | 7 | 1048 | |||
XM_011531901.1 | XP_011530203.1 | 8 | 1036 | |||
XM_017008130.1 | XP_016863619.1 | 8 | 1313 | |||
XM_017008131.1 | XP_016863620.1 | 7 | 1024 | |||
XM_017008132.1 | XP_016863621.1 | 8 | 1313 | |||
XM_017008133.1 | XP_016863622.1 | 7 | 1100 | |||
GenBank mRNA | AF020340.1 | - | 8 | 1005 | ||
AK296680.1 | - | 9 | 1221 | |||
AK299916.1 | - | 8 | 1034 | |||
AK300296.1 | - | 9 | 1140 | |||
AK307838.1 | - | 9 | 1250 | |||
AK315199.1 | - | 8 | 1075 | |||
AK315944.1 | - | 7 | 1029 | |||
BC047620.1 | - | 8 | 1036 | 5264160 | ||
X66533.1 | - | 8 | 1042 |