Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002806.3 | NP_002797.3 | 1, 2 | 131 | ||
XM_017021472.1 | XP_016876961.1 | 1, 2 | 108 | |||
GenBank mRNA | AF006305.1 | - | 1, 2 | 104 | ||
AK307087.1 | - | 1, 2 | 107 | |||
AK313670.1 | - | 1, 2 | 99 | |||
BC005390.1 | - | 1, 2 | 109 | 3996906 | ||
BM788029.1 | - | 1, 2 | 131 | |||
BT006843.1 | - | 1, 2 | 89 | |||
CR456709.1 | - | 1, 2 | 89 | |||
D78275.1 | - | 1, 2 | 105 | |||
FJ224299.1 | - | 1, 2 | 105 |